Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wipf1
Entrez Gene ID
117538
Gene Name
WAS/WASL interacting protein family, member 1
Gene Aliases
Waspip, Wip
Chromosome Location
Chr.3: 60150001 - 60207125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057192.2 NP_476540.2 4 - 5 1249 147
GenBank mRNA AJ303456.1 - 3 - 4 1082 147
BC072475.1 - 4 - 5 1249 147

View Full Product Details