Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Creb3l1
Entrez Gene ID
362165
Gene Name
cAMP responsive element binding protein 3-like 1
Gene Aliases
Oasis
Chromosome Location
Chr.3: 80892427 - 80933339 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005562.1 NP_001005562.1 7 - 8 1356 129
XM_006234584.1 XP_006234646.1 7 - 8 1412 129
GenBank mRNA BC081951.1 - 7 - 8 1356 129

View Full Product Details