The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014070.2 | NP_001014092.2 | 7 - 8 | 1284 | 98 |
XM_006239310.2 | XP_006239372.1 | 7 - 8 | 1540 | 98 | |
XM_006239311.2 | XP_006239373.1 | 7 - 8 | 2565 | 98 | |
XM_006239312.2 | XP_006239374.1 | 7 - 8 | 1352 | 98 | |
GenBank mRNA | BC089223.1 | - | 7 - 8 | 1284 | 98 |