Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo6
Entrez Gene ID
192351
Gene Name
F-box protein 6
Gene Aliases
Fbg2, Fbxo6b
Chromosome Location
Chr.5: 164956313 - 164962072 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239373.2 XP_006239435.1 1 - 2 227 81

View Full Product Details