Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mthfr
Entrez Gene ID
362657
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
Gene Aliases
-
Chromosome Location
Chr.5: 164844653 - 164864360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239413.2 XP_006239475.1 10 - 11 3236 97
XM_006239414.2 XP_006239476.1 10 - 11 1734 97
XM_006239416.1 XP_006239478.1 10 - 11 1901 97
XM_006239417.1 XP_006239479.1 10 - 11 1898 97
XM_006239418.1 XP_006239480.1 9 - 10 1833 97
XM_342975.6 XP_342976.3 10 - 11 3239 97

View Full Product Details