Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pusl1
Entrez Gene ID
362681
Gene Name
pseudouridylate synthase-like 1
Gene Aliases
-
Chromosome Location
Chr.5: 173336034 - 173339890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ220869.1 - 5 - 6 564 73

View Full Product Details