Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Capzb
Entrez Gene ID
298584
Gene Name
capping protein (actin filament) muscle Z-line, beta
Gene Aliases
-
Chromosome Location
Chr.5: 157642748 - 157742487 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005903.1 NP_001005903.1 7 - 8 738 74
XM_006239171.2 XP_006239233.1 8 - 9 1790 74
XM_006239172.2 XP_006239234.1 8 - 9 1787 74
XM_006239174.2 XP_006239236.1 7 - 8 1717 74
XM_006239175.2 XP_006239237.1 7 - 8 1713 74
XM_006239177.2 XP_006239239.1 7 - 8 767 74
GenBank mRNA BC083861.1 - 7 - 8 738 74

View Full Product Details