Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo42
Entrez Gene ID
362646
Gene Name
F-box protein 42
Gene Aliases
-
Chromosome Location
Chr.5: 159670662 - 159729070 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108691.1 NP_001102161.1 3 - 4 662 78
XM_006239253.2 XP_006239315.1 3 - 4 662 78

View Full Product Details