Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb8os
Entrez Gene ID
297885
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
RGD1562249, Zbtb8os-ps1
Chromosome Location
Chr.5: 147535585 - 147547252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750036.3 XP_003750084.3 4 - 5 760 89

View Full Product Details