Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnl2
Entrez Gene ID
362593
Gene Name
guanine nucleotide binding protein-like 2 (nucleolar)
Gene Aliases
RGD1305006
Chromosome Location
Chr.5: 143028031 - 143053418 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025736.1 NP_001020907.1 12 - 13 1480 73
GenBank mRNA BC099173.1 - 12 - 13 1480 73

View Full Product Details