Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl21
Entrez Gene ID
313743
Gene Name
kelch-like family member 21
Gene Aliases
RGD1305863
Chromosome Location
Chr.5: 169181418 - 169190073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107996.1 NP_001101466.1 1 - 2 1019 72

View Full Product Details