Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfrsf8
Entrez Gene ID
25069
Gene Name
tumor necrosis factor receptor superfamily, member 8
Gene Aliases
Cd30
Chromosome Location
Chr.5: 163186349 - 163232139 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019135.1 NP_062008.1 10 - 11 1079 89
GenBank mRNA D42117.1 - 10 - 11 1079 89

View Full Product Details