Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trappc3
Entrez Gene ID
362599
Gene Name
trafficking protein particle complex 3
Gene Aliases
-
Chromosome Location
Chr.5: 144281720 - 144295306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008376.1 NP_001008377.1 4 - 5 496 106
GenBank mRNA BC086377.1 - 4 - 5 496 106
FQ214189.1 - 4 - 5 552 106

View Full Product Details