Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hcrtr1
Entrez Gene ID
25593
Gene Name
hypocretin (orexin) receptor 1
Gene Aliases
Hctr1
Chromosome Location
Chr.5: 148346029 - 148355404 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013064.1 NP_037196.1 7 - 8 1532 86
GenBank mRNA AF041244.1 - 7 - 8 1532 86

View Full Product Details