Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nmnat1
Entrez Gene ID
298653
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.5: 166409460 - 166430254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037556.2 NP_001032645.2 1 - 2 38 72
GenBank mRNA AY788917.1 - 1 - 2 32 72
BC127446.1 - 1 - 2 38 72

View Full Product Details