Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttll10
Entrez Gene ID
298692
Gene Name
tubulin tyrosine ligase-like family, member 10
Gene Aliases
-
Chromosome Location
Chr.5: 173471020 - 173517785 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024758.2 NP_001019929.1 9 - 10 1052 106
NM_001170533.1 NP_001164004.1 10 - 11 1474 106
XM_006239559.2 XP_006239621.1 10 - 11 1634 106
XM_006239562.2 XP_006239624.1 8 - 9 1282 106
XM_008764356.1 XP_008762578.1 11 - 12 2921 106
XM_008764357.1 XP_008762579.1 10 - 11 1430 106
GenBank mRNA BC083833.1 - 7 - 8 891 106
BC097417.1 - 9 - 10 1052 106

View Full Product Details