The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017289.1 | NP_058985.1 | 8 - 9 | 1109 | 72 |
GenBank mRNA | BC087714.1 | - | 8 - 9 | 1074 | 72 |
M35162.1 | - | 8 - 9 | 1109 | 72 |