The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106743.1 | NP_001100213.1 | 11 - 12 | 2275 | 57 |
XM_006240323.2 | XP_006240385.1 | 11 - 12 | 2749 | 57 | |
XM_006240324.2 | XP_006240386.1 | 11 - 12 | 2745 | 57 | |
XM_006240325.2 | XP_006240387.1 | 11 - 12 | 2690 | 57 | |
XM_008764752.1 | XP_008762974.1 | 8 - 9 | 1662 | 57 | |
XM_008764753.1 | XP_008762975.1 | 8 - 9 | 1838 | 57 |