Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k9
Entrez Gene ID
500690
Gene Name
mitogen-activated protein kinase kinase kinase 9
Gene Aliases
RGD1562149
Chromosome Location
Chr.6: 105458867 - 105518971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100872.1 NP_001094342.1 2 - 3 829 67
XM_006240303.2 XP_006240365.1 2 - 3 1052 67

View Full Product Details