The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115034.1 | NP_001108506.1 | 2 - 3 | 406 | 88 |
GenBank mRNA | BC158698.1 | - | 2 - 3 | 363 | 88 |
FQ232113.1 | - | 2 - 3 | 368 | 88 | |
FQ233084.1 | - | 2 - 3 | 368 | 88 |