Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnal1
Entrez Gene ID
685664
Gene Name
dynein, axonemal, light chain 1
Gene Aliases
Dnalc1
Chromosome Location
Chr.6: 107596785 - 107623869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109477.2 NP_001102947.2 2 - 3 161 145
XM_006240352.2 XP_006240414.1 3 - 4 370 145
GenBank mRNA FQ211665.1 - - 165 145

View Full Product Details