Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tc2n
Entrez Gene ID
500707
Gene Name
tandem C2 domains, nuclear
Gene Aliases
Mtac2d1, Tac2-N
Chromosome Location
Chr.6: 125437767 - 125601408 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025152.1 NP_001020323.1 10 - 11 1141 148
XM_008764831.1 XP_008763053.1 9 - 10 2804 148
XM_008764832.1 XP_008763054.1 10 - 11 2453 148
XM_008764833.1 XP_008763055.1 10 - 11 2700 148
XM_008764834.1 XP_008763056.1 9 - 10 1674 148
XM_008764835.1 XP_008763057.1 10 - 11 1705 148
XM_008764836.1 XP_008763058.1 10 - 11 1323 148
XM_008764837.1 XP_008763059.1 10 - 11 1668 148
XM_008764838.1 XP_008763060.1 9 - 10 1637 148
XM_008764839.1 XP_008763061.1 9 - 10 1203 148
XM_008764840.1 XP_008763062.1 8 - 9 1070 148
GenBank mRNA BC088432.1 - 10 - 11 1141 148
FQ211170.1 - 10 - 11 1259 148

View Full Product Details