Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Calm1
Entrez Gene ID
24242
Gene Name
calmodulin 1
Gene Aliases
CaMI
Chromosome Location
Chr.6: 124217241 - 124225292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031969.2 NP_114175.1 2 - 3 143 80
GenBank mRNA AF178845.1 - - 78 80
FQ211760.1 - 2 - 3 210 80
FQ212721.1 - - 548 80
FQ213227.1 - 2 - 3 256 80
FQ225380.1 - 2 - 3 169 80
FQ226971.1 - 2 - 3 215 80
FQ227186.1 - 2 - 3 215 80
X13933.1 - 2 - 3 143 80

View Full Product Details