Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erh
Entrez Gene ID
681415
Gene Name
enhancer of rudimentary homolog (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.6: 104280390 - 104290905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109442.1 NP_001102912.1 1 - 2 344 59

View Full Product Details