Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erh
Entrez Gene ID
681415
Gene Name
enhancer of rudimentary homolog (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.6: 104280390 - 104290905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109442.1 NP_001102912.1 4 - 5 616 150
XM_006240290.2 XP_006240352.1 3 - 4 318 150
XM_008764848.1 XP_008763070.1 3 - 4 300 150
GenBank mRNA BC166864.1 - 3 - 4 233 150
FQ224144.1 - 3 - 4 323 150

View Full Product Details