The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108716.1 | NP_001102186.1 | 13 - 14 | 1427 | 65 |
XM_006240347.2 | XP_006240409.1 | 13 - 14 | 1430 | 65 | |
XM_008764814.1 | XP_008763036.1 | 12 - 13 | 1478 | 65 |