Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata7
Entrez Gene ID
192225
Gene Name
spermatogenesis associated 7
Gene Aliases
RSD-3, Wmp1
Chromosome Location
Chr.6: 122603252 - 122648718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138862.3 NP_620217.2 6 - 7 891 101
XM_006240413.2 XP_006240475.1 5 - 6 874 101
XM_006240414.2 XP_006240476.1 6 - 7 966 101
XM_006240415.2 XP_006240477.1 4 - 5 799 101
XM_008764737.1 XP_008762959.1 5 - 6 845 101
GenBank mRNA CK470849.1 - 2 - 3 555 101

View Full Product Details