The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108718.1 | NP_001102188.1 | 2 - 3 | 307 | 68 |
XM_006240394.2 | XP_006240456.1 | 2 - 3 | 313 | 68 |