Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itpk1
Entrez Gene ID
500709
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
-
Chromosome Location
Chr.6: 126448920 - 126582034 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191985.1 NP_001178914.1 4 - 5 366 69
XM_008764841.1 XP_008763063.1 3 - 4 1234 69
XM_008764842.1 XP_008763064.1 3 - 4 1705 69
XM_008764843.1 XP_008763065.1 3 - 4 4212 69

View Full Product Details