Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gar1
Entrez Gene ID
499709
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
Nola1
Chromosome Location
Chr.2: 235250453 - 235258460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024306.1 NP_001019477.1 2 - 3 486 126
XM_006232006.2 XP_006232068.1 2 - 3 1214 126
XM_008761519.1 XP_008759741.1 2 - 3 1215 126
GenBank mRNA BC079131.1 - 2 - 3 486 126

View Full Product Details