The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014043.1 | NP_001014065.1 | 5 - 6 | 1314 | 86 |
XM_006233311.2 | XP_006233373.1 | 6 - 7 | 2250 | 86 | |
XM_008761501.1 | XP_008759723.1 | 5 - 6 | 2166 | 86 | |
GenBank mRNA | BC085803.1 | - | 5 - 6 | 1314 | 86 |
DQ071571.2 | - | 4 - 5 | 885 | 86 |