Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssu72
Entrez Gene ID
298681
Gene Name
SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae)
Gene Aliases
RGD1307854
Chromosome Location
Chr.5: 173152964 - 173182720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025657.1 NP_001020828.1 4 - 5 743 81
GenBank mRNA BC099142.1 - 4 - 5 743 81

View Full Product Details