Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddost
Entrez Gene ID
313648
Gene Name
dolichyl-diphosphooligosaccharide-protein glycosyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 156668924 - 156676036 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012104.1 NP_001012104.1 1 - 2 165 73
GenBank mRNA BC082075.1 - 1 - 2 165 73
FQ228905.1 - 1 - 2 197 73
FQ229122.1 - 1 - 2 177 73

View Full Product Details