Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnq4
Entrez Gene ID
298496
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 4
Gene Aliases
-
Chromosome Location
Chr.5: 139625783 - 139677300 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764109.1 XP_008762331.1 9 - 10 1294 65
XM_008764111.1 XP_008762333.1 9 - 10 1019 65

View Full Product Details