Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem57
Entrez Gene ID
313618
Gene Name
transmembrane protein 57
Gene Aliases
-
Chromosome Location
Chr.5: 153122330 - 153184940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025699.2 NP_001020870.2 10 - 11 1970 75
XM_008764240.1 XP_008762462.1 9 - 10 2166 75
XM_008764241.1 XP_008762463.1 10 - 11 1922 75
GenBank mRNA AY845021.1 - 10 - 11 1970 75
BC097999.1 - 10 - 11 1896 75

View Full Product Details