Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bai2
Entrez Gene ID
313058
Gene Name
brain-specific angiogenesis inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.5: 148168516 - 148231587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107914.1 NP_001101384.1 16 - 17 2863 64
XM_006238934.1 XP_006238996.1 17 - 18 2971 64
XM_006238935.2 XP_006238997.1 17 - 18 3334 64
XM_006238936.1 XP_006238998.1 17 - 18 2971 64
XM_006238937.1 XP_006238999.1 17 - 18 2971 64
XM_006238938.1 XP_006239000.1 16 - 17 2806 64
XM_006238939.1 XP_006239001.1 16 - 17 2806 64
XM_008764141.1 XP_008762363.1 17 - 18 2971 64
XM_008764142.1 XP_008762364.1 16 - 17 2806 64
XM_008764143.1 XP_008762365.1 16 - 17 2806 64

View Full Product Details