Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai2
Entrez Gene ID
313058
Gene Name
brain-specific angiogenesis inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.5: 148168516 - 148231587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107914.1 NP_001101384.1 26 - 27 4378 84
XM_006238934.1 XP_006238996.1 27 - 28 4486 84
XM_006238935.2 XP_006238997.1 27 - 28 4849 84
XM_006238937.1 XP_006238999.1 26 - 27 4387 84
XM_006238938.1 XP_006239000.1 26 - 27 4321 84
XM_006238939.1 XP_006239001.1 26 - 27 4321 84
XM_008764142.1 XP_008762364.1 25 - 26 4222 84
XM_008764143.1 XP_008762365.1 25 - 26 4222 84

View Full Product Details