Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bai2
Entrez Gene ID
313058
Gene Name
brain-specific angiogenesis inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.5: 148168516 - 148231587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107914.1 NP_001101384.1 9 - 10 1973 87
XM_006238934.1 XP_006238996.1 10 - 11 2081 87
XM_006238935.2 XP_006238997.1 10 - 11 2444 87
XM_006238936.1 XP_006238998.1 10 - 11 2081 87
XM_006238937.1 XP_006238999.1 10 - 11 2081 87
XM_006238938.1 XP_006239000.1 9 - 10 1916 87
XM_006238939.1 XP_006239001.1 9 - 10 1916 87
XM_008764141.1 XP_008762363.1 10 - 11 2081 87
XM_008764142.1 XP_008762364.1 9 - 10 1916 87
XM_008764143.1 XP_008762365.1 9 - 10 1916 87

View Full Product Details