The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130405.1 | NP_569089.1 | 8 - 9 | 1318 | 127 |
GenBank mRNA | AF305619.1 | - | 8 - 9 | 1318 | 127 |
AF393783.1 | - | 8 - 9 | 1235 | 127 | |
BC061987.1 | - | 8 - 9 | 1342 | 127 |