Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Isg15
Entrez Gene ID
298693
Gene Name
ISG15 ubiquitin-like modifier
Gene Aliases
G1p2
Chromosome Location
Chr.5: 173624862 - 173629131 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106700.1 NP_001100170.1 1 - 2 146 82
XM_006239563.2 XP_006239625.1 2 - 3 740 82

View Full Product Details