Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epha10
Entrez Gene ID
298528
Gene Name
EPH receptor A10
Gene Aliases
RGD1562679
Chromosome Location
Chr.5: 142875683 - 142910537 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135709.1 NP_001129181.1 1 - 2 197 125
XM_006238858.2 XP_006238920.1 1 - 2 195 125

View Full Product Details