Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem39b
Entrez Gene ID
362608
Gene Name
transmembrane protein 39b
Gene Aliases
-
Chromosome Location
Chr.5: 147889874 - 147910057 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014192.1 NP_001014214.1 4 - 5 543 61
GenBank mRNA BC082025.1 - 4 - 5 543 61

View Full Product Details