The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006979.1 | NP_001006980.1 | 4 - 5 | 822 | 59 |
GenBank mRNA | BC083869.1 | - | 4 - 5 | 822 | 59 |