Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Necap2
Entrez Gene ID
298598
Gene Name
NECAP endocytosis associated 2
Gene Aliases
-
Chromosome Location
Chr.5: 159589856 - 159602258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199096.1 NP_954527.1 3 - 4 320 73
XM_006239179.2 XP_006239241.1 3 - 4 353 73
XM_006239180.2 XP_006239242.1 3 - 4 354 73
GenBank mRNA BC061826.1 - 3 - 4 320 73

View Full Product Details