Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sepn1
Entrez Gene ID
362624
Gene Name
selenoprotein N, 1
Gene Aliases
-
Chromosome Location
Chr.5: 152748497 - 152765498 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764190.1 XP_008762412.1 8 - 9 1236 66

View Full Product Details