Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tp73
Entrez Gene ID
362675
Gene Name
tumor protein p73
Gene Aliases
P73, Trp73
Chromosome Location
Chr.5: 171355876 - 171415354 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108696.1 NP_001102166.1 4 - 5 567 73

View Full Product Details