Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tp73
Entrez Gene ID
362675
Gene Name
tumor protein p73
Gene Aliases
P73, Trp73
Chromosome Location
Chr.5: 171355876 - 171415354 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108696.1 NP_001102166.1 5 - 6 758 64

View Full Product Details