Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem82
Entrez Gene ID
298605
Gene Name
transmembrane protein 82
Gene Aliases
RGD1566169
Chromosome Location
Chr.5: 160170617 - 160175141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239327.2 XP_006239389.1 2 - 3 799 106
XM_008764305.1 XP_008762527.1 2 - 3 799 106
XM_008764306.1 XP_008762528.1 2 - 3 799 106
XM_238429.8 XP_238429.5 2 - 3 799 106

View Full Product Details