Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pqlc2
Entrez Gene ID
362642
Gene Name
PQ loop repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 157744945 - 157759369 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239240.2 XP_006239302.1 1 - 2 84 79
XM_006239244.2 XP_006239306.1 1 - 2 84 79

View Full Product Details