The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007554.1 | NP_001007555.1 | 7 - 8 | 1011 | 64 |
XM_008764273.1 | XP_008762495.1 | 7 - 8 | 1778 | 64 | |
XM_008764274.1 | XP_008762496.1 | 6 - 7 | 941 | 64 | |
XM_008764275.1 | XP_008762497.1 | 7 - 8 | 1010 | 64 | |
XM_008764276.1 | XP_008762498.1 | 7 - 8 | 1161 | 64 | |
GenBank mRNA | BC086398.1 | - | 7 - 8 | 1011 | 64 |